Learn More
Aviva Systems Biology Corp BACH2 Antibody - middle region (ARP39513_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39513_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BACH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human BACH2. Alternate names: -ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53141