Learn More
Bachem H-Phe-2-chlorotrityl resin (200-400 mesh, 0.20-0.49 mmol/g)
SDP
Catalog No. 501947254
Encompass_Preferred
H-Phe-2-chlorotrityl resin (200-400 mesh, 0.20-0.49 mmol/g), 5g.
Catalog No. 50-194-7254 Supplier Bachem Supplier No. 4072916.0005
H-Phe-2-chlorotrityl resin (200-400 mesh, 0.20-0.49 mmol/g), 5g.