Learn More
Bachem H-Ser(tBu)-2-chlorotrityl resin (100-200 mesh, 0.50-0.90 mmol/g)
SDP
Catalog No. 501947308
Encompass_Preferred
H-Ser(tBu)-2-chlorotrityl resin (100-200 mesh, 0.50-0.90 mmol/g), 1g.
Catalog No. 50-194-7308 Supplier Bachem Supplier No. 4063685.0001
H-Ser(tBu)-2-chlorotrityl resin (100-200 mesh, 0.50-0.90 mmol/g), 1g.