Learn More
Bachem Fmoc-Ser(GlcNAc(Ac)3-b-D)-OH
SDP
Supplier:  Bachem 4042305.0100
Encompass_Preferred
Fmoc-Ser(GlcNAc(Ac)3-b-D)-OH, 160067-63-0, C32H36N2O13, Mr 656.64, 100mg. N-a-Fmoc-O-b-(2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-b-D-glucopyranosyl)-L-serine.
Catalog No. 50-194-7304