Learn More
Sigma Aldrich Fine Chemicals Biosciences BachiR glass oven for dry1EA
Catalog No. 502908778
Encompass
Small drying oven with metal housing and robust platform a transparent sample container suited for removing residual solvent up to a maximum temperature of 300C. Visual monitoring of the sample chamber graphic display with manual and programmable mode. Adjustable temperature and time control programmable for three steps..nbspRequires minimal space.nbspShorter heating time.nbspHigh energy efficiency.nbspSteady vacuum.nbspNo cross contamination.nbspOptimal visibility of the substance.nbspLow investment costs.nbspOption of keeping the sample in motion.nbspOutput to start and stop the vacuum pump.nbspWorking angle can be easily set from 0 to 90.nbspTemperature and time can be programmed at 3 levels.nbspVacuum-proof design.nbspOptional ventilation with inert gas.nbspNo cross contamination
Catalog No. 50-290-8778 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5630801EA
Add to Cart
Add to Cart
Request a Quote
Small drying oven with metal housing and robust platform a transparent sample container suited for removing residual solvent up to a maximum temperature of 300C. Visual monitoring of the sample chamber graphic display with manual and programmable mode. Adjustable temperature and time control programmable for three steps..nbspRequires minimal space.nbspShorter heating time.nbspHigh energy efficiency.nbspSteady vacuum.nbspNo cross contamination.nbspOptimal visibility of the substance.nbspLow investment costs.nbspOption of keeping the sample in motion.nbspOutput to start and stop the vacuum pump.nbspWorking angle can be easily set from 0 to 90.nbspTemperature and time can be programmed at 3 levels.nbspVacuum-proof design.nbspOptional ventilation with inert gas.nbspNo cross contamination