Learn More
ATCC Staphylococcus aureus; subsp. aureus
Supplier:  ATCC BAA39
Encompass
Multidrug-resistant bacterial strain that was isolated in 1993 from a human nose in Dunaujvaros, Hungary. This strain is pvl negative.
Catalog No. 50-237-9871