Learn More
ATCC Genomic DNA from Salmonella enterica; subsp. enterica; serovar Paratyphi B
Catalog No. 502380746
Encompass
Genomic DNA isolated from Salmonella enterica subsp. enterica serovar Paratyphi B strain SPB7. This bacterial strain is also available as ATCC Catalog No.: BAA-1250™.
Catalog No. 50-238-0746 Supplier ATCC Supplier No. BAA1250D5
Genomic DNA isolated from Salmonella enterica subsp. enterica serovar Paratyphi B strain SPB7. This bacterial strain is also available as ATCC Catalog No.: BAA-1250™.