Learn More
Novus Biologicals™ Bacillus anthracis protective antigen Antibody Blocking Peptide

Specifications
Specifications
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB120-13808 |
| Gene Alias | anthrax PA, Anthrax toxins translocating protein, PA, PA 83, PA83, PAG, Paga, Protective antigen |
| Quantity | 0.1 mg |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.