Learn More
ATCC Bacillus atrophaeus
Catalog No. 502384976
Encompass
Bacillus atrophaeus strain NRS 1221A is a whole-genome sequenced bacterial strain with applications as a test microorganism for sterilization and sterility assurance.
Catalog No. 50-238-4976 Supplier ATCC Supplier No. 9372
Bacillus atrophaeus strain NRS 1221A is a whole-genome sequenced bacterial strain with applications as a test microorganism for sterilization and sterility assurance.