Learn More
ATCC Bacillus cereus; 6 x 0.3mL
Catalog No. 502381969
Encompass
ATCC 10876-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 10876™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.
Catalog No. 50-238-1969 Supplier ATCC Supplier No. 10876MINIPACK
ATCC 10876-MINI-PACK™ consists of 6 ready-to-use vials of ATCC 10876™ frozen in 200 μL of glycerol stock, eliminating the need to rehydrate and culture the strain prior to use. Each vial is provided with a 2-D barcode for easy storage and tracking, as well as peel-off labels for fast and reliable recordkeeping.