Learn More
ATCC Pseudomonas putida; biotype b
Catalog No. 502380692
Encompass
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Pseudomonas putida strain MnB 1 is a whole-genome sequenced bacterium that was isolated from the manganese oxide crusts from a pipe. This strain oxidizes manganese.
Catalog No. 50-238-0692 Supplier ATCC Supplier No. 23483
Due to an upcoming ERP system upgrade, ATCC will have a temporary system shutdown from 11/20/26-12/1/26. All orders will experience a 5-6 days delay in processing. Orders will continue to be sent to ATCC during the blackout and will be held until 12/2/26 when they are released. Any questions, please reach out to Encompass.Supplierchannel@thermofisher.com.
Pseudomonas putida strain MnB 1 is a whole-genome sequenced bacterium that was isolated from the manganese oxide crusts from a pipe. This strain oxidizes manganese.