Learn More
TRIBIOSCIENCE INC Bacillus Coagulans qPCR Kit 100 Reactions No cross reactivity with other species.
SDP
Catalog No. NC3942564
Encompass_Preferred
Tribioscience’s Bacillus Coagulans qPCR Kit includes all components for bacillus coagulans amplification. The kit includes a qPCR super mix, the primer-probe mix labeled with FAM for the target gene, positive control, negative controls, and PCR internal controls labeled with Hex. This aid in a straightforward interpretation of the results. Applications - Detect bacillus-derived DNA in plant, cannabis, cannabis ingredients, grain, food, herbals, and animal feed. Key Features - High sensitivity and specificity for all bacillus coagulans species. High efficiency - the optimal systemic conditions for PCR amplifications. Streamlined protocol - just add DNA Template and water.
Catalog No. NC3942564 Supplier TRIBIOSCIENCE INC Supplier No. TBS42055100
Tribioscience’s Bacillus Coagulans qPCR Kit includes all components for bacillus coagulans amplification. The kit includes a qPCR super mix, the primer-probe mix labeled with FAM for the target gene, positive control, negative controls, and PCR internal controls labeled with Hex. This aid in a straightforward interpretation of the results. Applications - Detect bacillus-derived DNA in plant, cannabis, cannabis ingredients, grain, food, herbals, and animal feed. Key Features - High sensitivity and specificity for all bacillus coagulans species. High efficiency - the optimal systemic conditions for PCR amplifications. Streamlined protocol - just add DNA Template and water.