Learn More
ATCC Bacillus licheniformis
Catalog No. 502382186
Encompass
Bacillus licheniformis is a whole genome-sequenced bacterial type strain that has chitinase and chitobiase activity.
Catalog No. 50-238-2186 Supplier ATCC Supplier No. 14580
Bacillus licheniformis is a whole genome-sequenced bacterial type strain that has chitinase and chitobiase activity.