Learn More
ATCC Bacillus pacificus; NRS 248; genomic DNA
Catalog No. 502382830
Encompass
Genomic DNA isolated from Bacillus pacificus strain NRS 248 (ATCC 10987). This whole-genome sequenced product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-2830 Supplier ATCC Supplier No. 10987D5
Genomic DNA isolated from Bacillus pacificus strain NRS 248 (ATCC 10987). This whole-genome sequenced product can be used in PCR and other molecular biology applications.