Learn More
ATCC Bacillus pacificus
Catalog No. 502382814
Encompass
Bacillus pacificus strain NRS 248 is a xylose-positive variant. This bacterium is whole-genome sequenced and is a microbiome standard component.
Catalog No. 50-238-2814 Supplier ATCC Supplier No. 10987
Bacillus pacificus strain NRS 248 is a xylose-positive variant. This bacterium is whole-genome sequenced and is a microbiome standard component.