Learn More
ATCC Bacillus paralicheniformis
Catalog No. 502382024
Encompass
This strain of Bacillus paralicheniformis was isolated from a cassava tuber plant. This strain has applications as a quality control strain or in studies of proteinase.
Catalog No. 50-238-2024 Supplier ATCC Supplier No. 12759
This strain of Bacillus paralicheniformis was isolated from a cassava tuber plant. This strain has applications as a quality control strain or in studies of proteinase.