Learn More
ATCC Bacillus sp.
Catalog No. 502383988
Encompass
Bacillus sp. strain NCTC 8241 [CCM 2218, DSM 361, NCIB 8982] is a whole-genome sequenced bacterial strain. This bacterium has applications in assays of chlortetracycline, erythromycin, framycetin, gentamicin, lymecycline, neomycin, oxytetracycline, and tetracycline. It can also be used as a sterilization control.
Catalog No. 50-238-3988 Supplier ATCC Supplier No. 14884
Bacillus sp. strain NCTC 8241 [CCM 2218, DSM 361, NCIB 8982] is a whole-genome sequenced bacterial strain. This bacterium has applications in assays of chlortetracycline, erythromycin, framycetin, gentamicin, lymecycline, neomycin, oxytetracycline, and tetracycline. It can also be used as a sterilization control.