Learn More
TRIBIOSCIENCE INC Bacillus Species qPCR Kit, 100 Reactions
SDP
Catalog No. NC3454595
Encompass_Preferred
Tribioscience’s Bacillus Species qPCR Kit includes qPCR super mix, primer probe mix, Bacillus positive, negative controls. The target probe has been labeled with FAM for the target gene, and PCR internal control probe is labeled with Hex. These aid in a straightforward interpretation of the results.
Catalog No. NC3454595 Supplier TRIBIOSCIENCE INC Supplier No. TBS42034100
Tribioscience’s Bacillus Species qPCR Kit includes qPCR super mix, primer probe mix, Bacillus positive, negative controls. The target probe has been labeled with FAM for the target gene, and PCR internal control probe is labeled with Hex. These aid in a straightforward interpretation of the results.