Learn More
ATCC Pantoea stewartii; subsp. stewartii
Supplier:  ATCC 8199
Encompass
Major corn pathogen that causes Stewart's Bacterial Leaf Blight. This strain was isolated in 1941 from sweet corn var. Rugosa in Iowa, United States.
Catalog No. 50-238-0637