Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 4 (d4) 2mg
Catalog No. 502381497
Encompass
Purified pneumococcal polysaccharide type 4 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 4 is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 4 can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.
Catalog No. 50-238-1497 Supplier ATCC Supplier No. 34X
Purified pneumococcal polysaccharide type 4 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal powder type 4 is a component of the current 13-valent vaccine against Streptococcus pneumoniae. The pneumococcal polysaccharide type 4 can be used to assay for type-specific antibodies, research immune responses and study immunodeficiencies.