Learn More
ATCC Bacillus subtilis
Supplier:  ATCC 6051
Encompass
Bacillus subtilis strain Marburg is a whole-genome sequenced bacterial type strain that can be used in bacterial resistance testing, sterility assurance, and blood screening for phenylketonuria. This strain is also known to produce isoprene.
Catalog No. 50-238-3318