Learn More
TRIBIOSCIENCE INC Bacillus Subtilis qPCR Kit 100 Reactions
SDP
Catalog No. NC3942559
Encompass_Preferred
Tribioscience’s Bacillus Subtilis qPCR Kit includes bacillus subtilis positive and negative controls, PCR internal controls labeled with Hex, a qPCR super mix, and the primer-probe mix in which the probe has labeled with FAM for the target gene. These aid in a straightforward interpretation of the results. Applications - Detect bacillus-derived DNA in plant, cannabis, cannabis ingredients, grain, food, herbals, and animal feed. Key Features - High sensitivity and specificity for all bacillus subtilis species. High efficiency - the optimal systemic conditions for PCR amplifications. Streamlined protocol - just add DNA Template and water.
Catalog No. NC3942559 Supplier TRIBIOSCIENCE INC Supplier No. TBS42053100
Tribioscience’s Bacillus Subtilis qPCR Kit includes bacillus subtilis positive and negative controls, PCR internal controls labeled with Hex, a qPCR super mix, and the primer-probe mix in which the probe has labeled with FAM for the target gene. These aid in a straightforward interpretation of the results. Applications - Detect bacillus-derived DNA in plant, cannabis, cannabis ingredients, grain, food, herbals, and animal feed. Key Features - High sensitivity and specificity for all bacillus subtilis species. High efficiency - the optimal systemic conditions for PCR amplifications. Streamlined protocol - just add DNA Template and water.