Learn More
ATCC Providencia stuartii
Catalog No. 502381129
Encompass
Providencia stuartii strain 495 [CDC N1083] is a whole-genome sequenced bacterial strain. This culture can be used as a quality control strain for BBL products.
Catalog No. 50-238-1129 Supplier ATCC Supplier No. 33672
Add to Cart
Add to Cart
Providencia stuartii strain 495 [CDC N1083] is a whole-genome sequenced bacterial strain. This culture can be used as a quality control strain for BBL products.