Learn More
AdipoGen Bacimethrin
SDP
Supplier:  AdipoGen BVT0226C500
Encompass_Preferred
Chemical. CAS 3690-12-8. Formula C6H9N3O2. MW 155.2. Isolated from Streptomyces albus. Derivative of 5-Hydoxymethylcytosine Prod. No. BVT-0229. Antibiotic. Antibacterial. Thiamine antagonist. Phosphatase kinase inhibitor
Catalog No. 50-256-2057