Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bacitrac99, G-Biosciences
SCI_ED

Catalog No. S139751
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139751 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139751 Supplier G-Biosciences Supplier No. RC895
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bacitrac99 is the most pure form of bacitracin available.

Bacitrac99 is the most pure form of bacitracin available. Bacitracin is a cyclic polypeptide antibiotic isolated from Bacillus subtilis. It is made up of a variety of antimicrobial peptide fractions including A, A1, B, B1, B2, C, D, E, F, G, and X. This standard grade product is a mixture of these fractions, with bacitracin A exhibiting the most potent antimicrobial activity. Bacitracins B1 and B2 exhibit about 90% of the antimicrobial activity of bacitracin A when together.

Bacitracin targets the cell wall of gram-positive organisms such as Streptococcus pyogenes and Staphylococcus aureus. Bacitracin also functions as a useful tool to distinguish between group A and group B Beta-hemolytic Streptococcus species, which are susceptible and resistant, respectively.

Chemical Identifiers

Molecular Weight (g/mol) 1422.69

Specifications

Physical Form Powder
Quantity 10 g
Product Line Gibco
Percent Purity 0.99
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.