Learn More
Alkali Scientific Bacitracin, 25 G
SDP
Catalog No. 5000661068
Encompass_Preferred
Bacitracin is a polypeptide antibiotic used in laboratory research for its activity against Gram positive bacteria. It functions by interfering with bacterial cell wall synthesis, preventing proper cell membrane formation and leading to cell death. This makes it useful in microbiological applications where selective inhibition of Gram positive organisms is required. It is commonly used in culture media, contamination control, and antimicrobial studies. Bacitracin is also applied in cell culture systems to reduce unwanted bacterial growth. Supplied in a 25 gram quantity, it is suitable for routine laboratory use and preparation of antibiotic containing media.
Catalog No. 50-006-61068 Supplier Alkali Scientific Supplier No. BAC001CP
Bacitracin is a polypeptide antibiotic used in laboratory research for its activity against Gram positive bacteria. It functions by interfering with bacterial cell wall synthesis, preventing proper cell membrane formation and leading to cell death. This makes it useful in microbiological applications where selective inhibition of Gram positive organisms is required. It is commonly used in culture media, contamination control, and antimicrobial studies. Bacitracin is also applied in cell culture systems to reduce unwanted bacterial growth. Supplied in a 25 gram quantity, it is suitable for routine laboratory use and preparation of antibiotic containing media.