Learn More
Selleck Chemical LLC Bacitracin S4146-50mg
SDP
Catalog No. 503108379
Encompass_Preferred
Bacitracin is a mixture of related cyclic polypeptides produced by organisms of the licheniformis group of Bacillus subtilis var Tracy which disrupts both gram positive and gram negative bacteria by interfering with cell wall and peptidoglycan synthesis
Catalog No. 50-310-8379 Supplier Selleck Chemical LLC Supplier No. S414650MG
Bacitracin is a mixture of related cyclic polypeptides produced by organisms of the licheniformis group of Bacillus subtilis var Tracy which disrupts both gram positive and gram negative bacteria by interfering with cell wall and peptidoglycan synthesis