Learn More
Gold Biotechnology Inc Bacitracin, USP Grade 1 g
SDP
Catalog No. NC2014279
Encompass
Bacitracin is a cyclic polypeptide antibiotic isolated from Bacillus subtilis. It is made up of a variety of antimicrobial peptide fractions including A, A1, B, B1, B2, C, D, E, F, G, and X. This standard grade product is a mixture of these fractions, with bacitracin A exhibiting the most potent antimicrobial activity. Bacitracins B1 and B2 exhibit about 90% of the antimicrobial activity of bacitracin A when together. Other forms of bacitracin have not been studied extensively, but may have negligible antimicrobial properties. Bacitracin is freely soluble in aqueous solution.
Catalog No. NC2014279 Supplier Gold Biotechnology Inc Supplier No. B0701
Bacitracin is a cyclic polypeptide antibiotic isolated from Bacillus subtilis. It is made up of a variety of antimicrobial peptide fractions including A, A1, B, B1, B2, C, D, E, F, G, and X. This standard grade product is a mixture of these fractions, with bacitracin A exhibiting the most potent antimicrobial activity. Bacitracins B1 and B2 exhibit about 90% of the antimicrobial activity of bacitracin A when together. Other forms of bacitracin have not been studied extensively, but may have negligible antimicrobial properties. Bacitracin is freely soluble in aqueous solution.