Learn More
Selleck Chemical LLC Bacitracin Zinc S1791-50mg
SDP
Catalog No. 503109789
Encompass_Preferred
Bacitracin(Zinc Bacitracin) is an antibiotic that interferes with the dephosphorylation of C55-isoprenyl pyrophosphate a membrane carrier molecule that transports the building-blocks of the peptidoglycan bacterial cell wall outside of the inner membrane It inhibits cleavage of Tyr from Met-enkephalin with IC50 of 10 M
Catalog No. 50-310-9789 Supplier Selleck Chemical LLC Supplier No. S179150MG
Bacitracin(Zinc Bacitracin) is an antibiotic that interferes with the dephosphorylation of C55-isoprenyl pyrophosphate a membrane carrier molecule that transports the building-blocks of the peptidoglycan bacterial cell wall outside of the inner membrane It inhibits cleavage of Tyr from Met-enkephalin with IC50 of 10 M