Learn More
Apexbio Technology LLC Bacitracin Zinc 1405-89-6 50mg
SDP
Catalog No. 502240807
Encompass_Preferred
Bacitracin Zinc (CAS 1405-89-6) is a peptide antibiotic targeting the dephosphorylation of C55-isoprenyl pyrophosphate It is designed to inhibit this step thereby disturbing bacterial cell wall biosynthesis Bacitracin Zinc exerts its biological activity primarily by interfering with the dephosphorylation process It also inhibits the enzymatic cleavage of tyrosine from Met-enkephalin with an IC50 value of approximately 10 M Based on these pharmacological properties Bacitracin Zinc holds research potential in studying bacterial cell envelope biosynthesis enzymatic processes involving peptide substrates and enzyme assay development in biomedical investigations
This item is not returnable. View return policy
Catalog No. 50-224-0807 Supplier Apexbio Technology LLC Supplier No. B167150
Bacitracin Zinc (CAS 1405-89-6) is a peptide antibiotic targeting the dephosphorylation of C55-isoprenyl pyrophosphate It is designed to inhibit this step thereby disturbing bacterial cell wall biosynthesis Bacitracin Zinc exerts its biological activity primarily by interfering with the dephosphorylation process It also inhibits the enzymatic cleavage of tyrosine from Met-enkephalin with an IC50 value of approximately 10 M Based on these pharmacological properties Bacitracin Zinc holds research potential in studying bacterial cell envelope biosynthesis enzymatic processes involving peptide substrates and enzyme assay development in biomedical investigations