Learn More
BINDER™ Back Access Port
SDP
Catalog No. 5000217533 Shop All BINDER Inc. Products
Click to view available options
Diameter (Metric) Inside:
30 mm
Catalog No. Diameter (Metric) Inside
50-002-17533 30 mm
1 options
Catalog No. 50-002-17533 Supplier BINDER™ Supplier No. 80121390
For Use With: BINDER™ units Series CB
  • For use in back
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
For Use With (Equipment) BINDER Units Series CB
Type Access Port
For Use With (Application) Back
Includes Silicone Plug
Diameter (Metric) Inside 30 mm