Learn More
BINDER™ Back Access Port (10 mm)
SDP
Catalog No. 08111508 Shop All BINDER Inc. Products
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers BF 400
Catalog No. For Use With (Equipment)
08-111-508 BINDER Chambers BF 400
1 options
Catalog No. 08-111-508 Supplier BINDER™ Supplier No. 80121281
Add to Cart
Add to Cart
For Use With: BF 400
  • For use in back
  • Includes silicone plug
  • For use with a wide range of BINDER™ incubators and chambers
Specifications
Description Access port with silicone plug - 10 mm, back
For Use With (Equipment) BINDER Chambers BF 400
Material Steel
Type Access Port
Includes Silicone plug
Diameter (Metric) Inside 10 mm