Learn More
Grainger Back Chamfer Milling Cutter 1/2 Carbide
Catalog No. 502768561
Encompass_Preferred
The Micro 100 Indexable Chamfering Tool is a high-performance chamfering tool designed for use on CNC lathes and machining centers. It features a carbide insert with four cutting edges for long tool life. The tool has a shank diameter of 1" and an insert size of 16mm. It can produce chamfers from .04" to .24" on steel, stainless steel, cast iron, aluminum and other materials. The patented insert geometry allows high feed rates and smooth cutting action. Made in USA by Micro 100, this chamfering tool is ideal for production chamfering applications.
Catalog No. 50-276-8561 Supplier Grainger Supplier No. 16T456
The Micro 100 Indexable Chamfering Tool is a high-performance chamfering tool designed for use on CNC lathes and machining centers. It features a carbide insert with four cutting edges for long tool life. The tool has a shank diameter of 1" and an insert size of 16mm. It can produce chamfers from .04" to .24" on steel, stainless steel, cast iron, aluminum and other materials. The patented insert geometry allows high feed rates and smooth cutting action. Made in USA by Micro 100, this chamfering tool is ideal for production chamfering applications.