Learn More
Grainger Back Chamfer Milling Cutter 3/8 Carbide
Catalog No. 502766029
Encompass_Preferred
The Micro 100 Indexable Chamfering Tool is a high-performance chamfering tool designed for use on CNC lathes and machining centers. It features a 16mm shank with a 45 degree lead angle insert for chamfering internal and external diameters. The tool comes with 5 IC908 carbide inserts with TiAlN coating for wear resistance and a clamping screw to hold the inserts in place. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. The tool provides an efficient method for putting a 45 degree chamfer on parts coming off the lathe or mill.
Catalog No. 50-276-6029 Supplier Grainger Supplier No. 16T455
The Micro 100 Indexable Chamfering Tool is a high-performance chamfering tool designed for use on CNC lathes and machining centers. It features a 16mm shank with a 45 degree lead angle insert for chamfering internal and external diameters. The tool comes with 5 IC908 carbide inserts with TiAlN coating for wear resistance and a clamping screw to hold the inserts in place. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. The tool provides an efficient method for putting a 45 degree chamfer on parts coming off the lathe or mill.