Learn More
Grainger Back Chamfering Tool
Catalog No. 502764882
Encompass_Preferred
The Micro 100 Indexable Chamfering Tool is a high-performance chamfering tool designed for chamfering, facing, and grooving operations. It features a carbide insert with six cutting edges for extended tool life. The tool has a shank diameter of 3/4 inches and an overall length of 4 inches. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. The chamfering tool provides excellent chip control and consistent chamfer quality in one pass. It is ideally suited for chamfering holes, slots, keyways, and more in machining applications. The tool requires a holder for mounting and comes with assembled screws for easy insert changes.
Catalog No. 50-276-4882 Supplier Grainger Supplier No. 16T463
The Micro 100 Indexable Chamfering Tool is a high-performance chamfering tool designed for chamfering, facing, and grooving operations. It features a carbide insert with six cutting edges for extended tool life. The tool has a shank diameter of 3/4 inches and an overall length of 4 inches. It can machine a variety of materials including steel, stainless steel, cast iron, aluminum, and more. The chamfering tool provides excellent chip control and consistent chamfer quality in one pass. It is ideally suited for chamfering holes, slots, keyways, and more in machining applications. The tool requires a holder for mounting and comes with assembled screws for easy insert changes.