Learn More
Grainger Back Chamfering Tool
Catalog No. 502768515
Encompass_Preferred
The Back Chamfering Tool 16T460 is a high speed steel chamfering tool designed for back chamfering operations. It features a 16mm shank diameter, 460mm overall length, and 45 degree chamfering angle. The tool has 6 flutes and is made of M2 grade high speed steel for wear resistance and durability. It can chamfer various materials like steel, stainless steel, cast iron, and non-ferrous metals. The tool provides efficient back chamfering in turning applications.
Catalog No. 50-276-8515 Supplier Grainger Supplier No. 16T460
The Back Chamfering Tool 16T460 is a high speed steel chamfering tool designed for back chamfering operations. It features a 16mm shank diameter, 460mm overall length, and 45 degree chamfering angle. The tool has 6 flutes and is made of M2 grade high speed steel for wear resistance and durability. It can chamfer various materials like steel, stainless steel, cast iron, and non-ferrous metals. The tool provides efficient back chamfering in turning applications.