Learn More
Minuteman™ Back Pack HEPA Vacuum Accessories
SDP
Catalog No. 19163084 Shop All Minuteman International, Inc. Products
Click to view available options
Product Type:
Pre-filter Disk
Vacuum Bag
Vacuum Filter
Material:
Cloth
Paper
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material
19-163-084 Vacuum Filter -
19-163-082 Pre-filter Disk -
19-163-081 Vacuum Bag Cloth
19-163-083 Vacuum Bag Paper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-163-084 Supplier Minuteman™ Supplier No. 293086
Use with the Minuteman Back Pack HEPA Vacuum
Vacuum provides trapping efficiency of 99.7% at 0.3μ.
Specifications
Product Type Vacuum Filter
Quantity Each
For Use With (Equipment) BVP Back Pack HEPA Vacuum
Filter Type HEPA
Efficiency 99.7%