Learn More
MSC Honeywell Lanyard, BackBiter Twin-Leg, 6 ft, Tie-Back, 1 ea
Catalog No. 502370327
Encompass
This shock absorbing lanyard is an all-in-one product featuring a SoftStop shock absorber with a specially-woven inner core that expands to reduce fall arrest forces. It also includes a cross-arm anchorage connector. The lanyard is made of heavy-duty polyester webbing and Cordura nylon webbing.
  • SoftStop shock absorber reduces fall arrest forces
  • Cross-arm anchorage connector
  • Heavy-duty polyester webbing construction
  • Cordura nylon webbing for five times more abrasion resistance
  • Includes 1 locking snap hook and 2 Miller 5K locking snap hooks
  • Provides top-to-bottom tie-back
  • Suitable for construction, confined space, roofing, manufacturing, transportation, steel, and metals industries
Catalog No. 50-237-0327 Supplier MSC Supplier No. 08684680
This shock absorbing lanyard is an all-in-one product featuring a SoftStop shock absorber with a specially-woven inner core that expands to reduce fall arrest forces. It also includes a cross-arm anchorage connector. The lanyard is made of heavy-duty polyester webbing and Cordura nylon webbing.
  • SoftStop shock absorber reduces fall arrest forces
  • Cross-arm anchorage connector
  • Heavy-duty polyester webbing construction
  • Cordura nylon webbing for five times more abrasion resistance
  • Includes 1 locking snap hook and 2 Miller 5K locking snap hooks
  • Provides top-to-bottom tie-back
  • Suitable for construction, confined space, roofing, manufacturing, transportation, steel, and metals industries