Learn More
Diagnostic Biosystems BACKGROUND BLOCKER Background Blocker (1X)
Catalog No. NC2049157
Encompass_Preferred
To block nonspecific background staining observed during immunostaining.\n\nBackground Blocker is unique serum-free blocking solution used to reduce nonspecific background staining. It is a universal blocking agent which eliminates the need to match species with the link antibody. This blocking reagent has been shown to be more superior than serum-based blocking solutions. In addition to immunihistochemistry, Background Blocker can also be used for ELISA and Western Blot techniques.
Catalog No. NC2049157 Supplier Diagnostic Biosystems Supplier No. K023
To block nonspecific background staining observed during immunostaining.\n\nBackground Blocker is unique serum-free blocking solution used to reduce nonspecific background staining. It is a universal blocking agent which eliminates the need to match species with the link antibody. This blocking reagent has been shown to be more superior than serum-based blocking solutions. In addition to immunihistochemistry, Background Blocker can also be used for ELISA and Western Blot techniques.