Learn More
INNOVEX BIOSCIENCES BACKGROUND BUSTER READY-TO-USE 1 LITER
SDP
Catalog No. 50486804
Encompass_Preferred
Number of slides: 11,200-16,000; Shelf life: 3 years; Peptide Blocker delivers complete eradication of background staining of animal, human tissues & cells stained by IHC, IF, in situ ( frozen, paraffin) & free floating sections; Short incubation step. A must for Mouse-on-Mouse staining.
Catalog No. 50-486-804 Supplier INNOVEX BIOSCIENCES Supplier No. NB3061L
Number of slides: 11,200-16,000; Shelf life: 3 years; Peptide Blocker delivers complete eradication of background staining of animal, human tissues & cells stained by IHC, IF, in situ ( frozen, paraffin) & free floating sections; Short incubation step. A must for Mouse-on-Mouse staining.