Learn More
INNOVEX BIOSCIENCES BACKGROUND BUSTER READY-TO-USE 125 ML
SDP
Catalog No. 50486805
Encompass_Preferred
Peptide Blocker delivers complete eradication of background staining of animal, human tissues & cells stained by IHC, IF, in situ ( frozen, paraffin) & free floating sections; Short incubation step. A must for Mouse-on-Mouse staining
Catalog No. 50-486-805 Supplier INNOVEX BIOSCIENCES Supplier No. NB306
Peptide Blocker delivers complete eradication of background staining of animal, human tissues & cells stained by IHC, IF, in situ ( frozen, paraffin) & free floating sections; Short incubation step. A must for Mouse-on-Mouse staining