Learn More
Biocare Medical, LLC Background Punisher - 100 ml
SDP
Catalog No. 5082380
Encompass_Preferred
Background Punisher is a universal blocking reagent used for reducing nonspecific background staining often found with immunohistochemistry. Background Punisher is specifically formulated for superior pH stability and is sodium azide and thimerosal free. It can be used with automated or manual staining protocols.
Catalog No. 50-823-80 Supplier Biocare Medical, LLC Supplier No. BP974L
Background Punisher is a universal blocking reagent used for reducing nonspecific background staining often found with immunohistochemistry. Background Punisher is specifically formulated for superior pH stability and is sodium azide and thimerosal free. It can be used with automated or manual staining protocols.