Learn More
Stoelting™ Backhaus Towel Clamp
SDP
Catalog No. 10000484 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
3.6 in.
5.25 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
10-000-484 3.6 in.
10-001-398 5.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-000-484 Supplier Stoelting™ Supplier No. 5212280P
Add to Cart
Add to Cart
Backhaus Towel Clamp is for fixing drapes to the skin of anesthetized patients.
Instrument has scissor action with ratchet fixation at the finger loops and sharp, incurving, needle-like blades.
Specifications
For Use With (Application) Neuroscience
Size 3.6 in.