Learn More
Med Vet International Miltex Vantage Backhaus Towel Clamp, 3.5in.
Catalog No. 502829586
Encompass_Preferred
The Miltex Vantage V97-504 Backhaus towel clamp is made from high quality Japanese stainless steel and measures 3.5" in length. It has a finger ring handle with a ratchet lock.
Catalog No. 50-282-9586 Supplier Med Vet International Supplier No. MILV97504
The Miltex Vantage V97-504 Backhaus towel clamp is made from high quality Japanese stainless steel and measures 3.5" in length. It has a finger ring handle with a ratchet lock.