Learn More
Sklar Instruments Clamp, Surgi-OR Backhaus Towel Clamp, Sklar 5-1/4 in., Curved, Perforated, 1 x 1 Prong, Stainless Steel, EA.
SDP
Catalog No. 501182911 Shop All Sklar Instruments Products
Encompass_Preferred
Clamp, Surgi-OR Backhaus Towel Clamp, Sklar, 5-1/4 in.,, Curved, Perforated, 1 x 1 Prong, Stainless Steel, EA. Surgi-OR Backhaus Towel Clamps are a perforating clamp used for grasping tissue, securing towels or drapes and holding or reducing small bone fractures. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is curved with 1X1 perforating prongs and 5-1/4 inches in length. Lifetime warranty against all manufacturer defects.
Catalog No. 50-118-2911 Supplier Sklar Instruments Supplier No. 95611
Clamp, Surgi-OR Backhaus Towel Clamp, Sklar, 5-1/4 in.,, Curved, Perforated, 1 x 1 Prong, Stainless Steel, EA. Surgi-OR Backhaus Towel Clamps are a perforating clamp used for grasping tissue, securing towels or drapes and holding or reducing small bone fractures. Hand-crafted in Pakistan, Surgi-OR mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is curved with 1X1 perforating prongs and 5-1/4 inches in length. Lifetime warranty against all manufacturer defects.