Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILC Dover Backpack for Sentinel XL

Catalog No. 19631064
Change view
Click to view available options
For Use With: Sentinel XL Blower
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-631-064 For Use With: Sentinel XL Blower
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-631-064 Supplier ILC Dover Supplier No. S2008
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sentinel XL PAPR backpack holds blowers and battery packs to keep hands free.

  • For use with Sentinel XL PAPR

Specifications

Color Black
Disposable Reusable
Product Type Backpack
For Use With (Equipment) Sentinel XL PAPR
Model S-2008
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.