Learn More
MSC Arborjet AccuFlo ISD Roller Tank Sprayer, 9 gal., 1 ea
Catalog No. 502817930
Encompass
This polyethylene tank sprayer is designed for various outdoor applications, including lawn, garden, and tree care. It features a durable construction and is suitable for use with various chemicals.
  • Polyethylene tank
  • Volume capacity of 9 gal.
  • Chemical safe
  • In-tank filter
  • Aluminum and brass wand
  • Brass nozzle
  • Synthetic rubber seal/gasket
  • Includes nozzle, shoulder strap, battery, wand, charger, adjustable nozzle
  • Spray patterns: cone, flat, stream
Catalog No. 50-281-7930 Supplier MSC Supplier No. 11669181
This polyethylene tank sprayer is designed for various outdoor applications, including lawn, garden, and tree care. It features a durable construction and is suitable for use with various chemicals.
  • Polyethylene tank
  • Volume capacity of 9 gal.
  • Chemical safe
  • In-tank filter
  • Aluminum and brass wand
  • Brass nozzle
  • Synthetic rubber seal/gasket
  • Includes nozzle, shoulder strap, battery, wand, charger, adjustable nozzle
  • Spray patterns: cone, flat, stream