Learn More
MSC Solo Backpack Sprayer, 4 Gallon, Piston, HDPE
Catalog No. 502837973
Encompass
The Solo 425 is a professional-grade backpack sprayer, ergonomically designed for comfort and durability. Its high-performance piston pump efficiently handles liquid formulations for various spraying tasks, ensuring a long service life with minimal maintenance.
  • Professional-grade design ensures durability and longevity.
  • Ergonomic design contours to the back for enhanced user comfort.
  • High-performance piston pump provides up to 90 psi for versatile liquid applications.
  • Large filling aperture simplifies tank filling and cleaning.
  • 28-inch unbreakable wand and commercial shut-off valve offer extended reach and reduce user fatigue.
  • High-quality seals resist a wide range of agricultural and household chemicals.
  • Includes four nozzles (adjustable, fan, hollow cone, jet stream) for varied spray patterns.
Catalog No. 50-283-7973 Supplier MSC Supplier No. 72905862
The Solo 425 is a professional-grade backpack sprayer, ergonomically designed for comfort and durability. Its high-performance piston pump efficiently handles liquid formulations for various spraying tasks, ensuring a long service life with minimal maintenance.
  • Professional-grade design ensures durability and longevity.
  • Ergonomic design contours to the back for enhanced user comfort.
  • High-performance piston pump provides up to 90 psi for versatile liquid applications.
  • Large filling aperture simplifies tank filling and cleaning.
  • 28-inch unbreakable wand and commercial shut-off valve offer extended reach and reduce user fatigue.
  • High-quality seals resist a wide range of agricultural and household chemicals.
  • Includes four nozzles (adjustable, fan, hollow cone, jet stream) for varied spray patterns.