Learn More
MSC Clarke Backpack Vacuum, Comfort Pak 10, HEPA, 1 ea
Catalog No. 502868759
Encompass
The Clarke Comfort Pak 10 is a backpack vacuum designed for efficient cleaning of various surfaces in commercial settings. It offers powerful suction and maneuverability for enhanced productivity.
  • Powerful 1300 W motor with 144 cfm airflow and 99 in waterlift suction
  • Features 10 quart capacity for extended cleaning tasks
  • Equipped with 4-stage filtration system and optional HEPA capability
  • Operates quietly at 62 dB A for comfortable use
  • Long 50 ft power cord for wide reach
  • Includes a comprehensive tool kit for versatile cleaning
  • Durable ABS plastic construction for longevity
Catalog No. 50-286-8759 Supplier MSC Supplier No. 35702935
The Clarke Comfort Pak 10 is a backpack vacuum designed for efficient cleaning of various surfaces in commercial settings. It offers powerful suction and maneuverability for enhanced productivity.
  • Powerful 1300 W motor with 144 cfm airflow and 99 in waterlift suction
  • Features 10 quart capacity for extended cleaning tasks
  • Equipped with 4-stage filtration system and optional HEPA capability
  • Operates quietly at 62 dB A for comfortable use
  • Long 50 ft power cord for wide reach
  • Includes a comprehensive tool kit for versatile cleaning
  • Durable ABS plastic construction for longevity