Learn More
DSI LLC Backpressure Regulator, variable 15 - 300psi, max. 50C, complete, for liquids only
Supplier:  DSI LLC JRBPR1
Encompass_Preferred
Backpressure Regulator, variable 15 - 300psi, max. 50C, complete, for liquids only
Catalog No. 50-225-5018